Learn More
Wnt-C59, MedChemExpress
Product Code. 16456728
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16456728 10 mg 10mg
16472807 50 mg 50mg
16469277 5 mg 5mg
16456866 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16456728 Supplier Medchem Express Supplier No. HY15659
MedChemExpress Wnt-C59 (C59) is a highly potent and oral porcupine (PORCN) inhibitor with an IC50 of 74 pM.
Wnt-C59 (C59) is a highly potent and oral porcupine (PORCN) inhibitor with an IC50 of 74 pM.
Specifications
Chemical Name or Material Wnt-C59
CAS 1243243-89-1
Color White
Purity Grade Notes Research
Molecular Formula C25H21N3O
Quantity 10 mg
Synonym C59
Solubility Information DMSO : 50 mg/mL (131.77 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C2=CC=CN=C2)C=C1)CC3=CC=C(C4=CC(C)=NC=C4)C=C3
Molecular Weight (g/mol) 379.45
Formula Weight 379.45
Percent Purity 99.32%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death