missing translation for 'onlineSavingsMsg'
Learn More

WM-8014, MedChemExpress

Product Code. 16426447
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16426447 5 mg 5mg
16484776 100 mg 100mg
16452836 25 mg 25mg
16433708 10 mg 10mg
16400097 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16426447 Supplier Medchem Express Supplier No. HY102060

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WM-8014 is an inhibitor of MOZ, a member of histone acetyltransferases, with an IC50 of 55 nM.

WM-8014 is an inhibitor of MOZ, a member of histone acetyltransferases, with an IC50 of 55 nM.

Specifications

Chemical Name or Material WM-8014
CAS 2055397-18-5
Color White
Purity Grade Notes Research
Molecular Formula C20H17FN2O3S
Quantity 5 mg
Solubility Information DMSO : ≥ 250 mg/mL (650.33 mM)
SMILES O=C(C1=CC(C2=CC=CC=C2)=CC(C)=C1F)NNS(=O)(C3=CC=CC=C3)=O
Molecular Weight (g/mol) 384.42
Formula Weight 384.42
Percent Purity 98.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.