Learn More
Benchmark Scientific™ Wire Shelf for UV-Clave
Click to view available options
For Use With (Equipment):
UV-Clave
Unit Size:
Each
Description
- For use in Benchmark UV-Clave
- Wire shelf
Specifications
Specifications
| Dimensions (L x W) | 8 x 10 in. |
| For Use With (Application) | Hold Items in UV-Clave |
| For Use With (Equipment) | UV-Clave |
| Material | Stainless Steel |