Learn More
Nordvlies WIPEX microfibre wipes ECONOMY, 240 g/sqm, 40x40 cm, blue
Brand:  Nordvlies 88002
WIPEX microfibre wipes ECONOMY, 240 g/sqm, 40x40 cm, blue
Product Code. 15721067
  • ONLINE EXCLUSIVE PRICE
    64.00 EUR
    46.48 EUR / Pack of 20
    Save 17.52 EUR
    27% Off