missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wilkinson’s Catalyst Sulfate Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Wilkinson’s Catalyst Sulfate Sodium Salt |
| Analyte or Component Names | Wilkinson’s Catalyst Sulfate Sodium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C54 H36 Cl Na9 O27 P3 Rh S9 |
| Synonym | Chlorotris(triphenylphosphine)rhodium Sulfate Sodium Salt, Rhodium tris(triphenylphosphine) Chloride Sulfate Sodium Salt, Tris(triphenylphosphine)chlororhodium Sulfate Sodium Salt, Tris(triphenylphosphine)rhodium(I) Chloride Sulfate Sodium Salt |
| InChI Formula | InChI=1S/3C18H15O9PS3.ClH.9Na.Rh/c3*19-29(20,21)16-7-1-13(2-8-16)28(14-3-9-17(10-4-14)30(22,23)24)15-5-11-18(12-6-15)31(25,26)27;;;;;;;;;;;/h3*1-12H,(H,19,20,21)(H,22,23,24)(H,25,26,27);1H;;;;;;;;;;/q;;;;10*+1/p-10 |
| Solution Type | Neat |
| SMILES | [Na]OS(=O)(=O)c1ccc(cc1)[P](c2ccc(cc2)S(=O)(=O)O[Na])(c3ccc(cc3)S(=O)(=O)O[Na])[Rh](Cl)([P](c4ccc(cc4)S(=O)(=O)O[Na])(c5ccc(cc5)S(=O)(=O)O[Na])c6ccc(cc6)S(=O)(=O)O[Na])[P](c7ccc(cc7)S(=O)(=O)O[Na])(c8ccc(cc8)S(=O)(=O)O[Na])c9ccc(cc9)S(=O)(=O)O[Na] |
| IUPAC Name | chloro-tris[tris(4-sodiooxysulfonylphenyl)-λ4-phosphanyl]rhodium |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?