Learn More
Cole-Parmer™ Wildco 1510-C20 Kemmerer Water Sampler
Shop All Cole Parmer Products
Click to view available options
Capacity (Metric):
1200 mL
2.2 L
Unit Size:
Each
Description
- Messenger activates the unique trip heads that ensure closure in fast flowing streams or turbulent waters, regardless of line angle
- Order messenger and line separately
- Includes a plastic carrying case
Specifications
Specifications
| Product Type | Water Sampler |
| Includes | Plastic carrying case |
| Closure Material | Silicone |
| Tube Material | Acrylic |
| Capacity (Metric) | 2.2 L |
| Style | Kemmerer Water Sampler |
| For Use With (Application) | Water sampling at depths of 3 to 600 ft. |