Learn More
Thermo Scientific Chemicals Wig-L-Bug™ Mixing/Grinding Mill Accessory, Ball Pestles, 9.52mm Dia.
Product Code. 11386119
Click to view available options
Item Description:
Quantity: 100/Pk.
Quantity: 25/Pk.
Unit Size:
Pack of 100
Pack of 25
Product Code. 11386119
Brand: Thermo Scientific Chemicals 042631.KM
Request Bulk or Custom Format
For use with Wig-L-Bug Mixing/Grinding Mill (AA42627KT)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Item Description Quantity: 25/Pk.
Diameter (English) Outer 0.375 in.
Diameter (Metric) Outer 9.52mm
Product Type Ball Pestle
For Use With (Equipment) Wig-L-Bug Mixing/Grinding Mill (AA42627KT)