Learn More
Motic™ Widefield Eyepiece WF10X/18mm
Click to view available options
Magnification Power:
10X
Unit Size:
Each
Description
- Widefield style provides improved field of view and greater eye relief.
- Magnification: 10X.
- Field of view: 18mm.
Specifications
Specifications
| Type | Widefield |
| Description | FOV: 18mm |
| For Use With (Equipment) | RED100-Series, Series 18, Series 28, PM Series, DM1802-A Microscopes |
| Magnification Power | 10X |