Learn More
Motic™ Widefield Eyepiece WF10X/18mm
Product Code. 15425102
Click to view available options
Magnification Power:
10X
Unit Size:
Each
Product Code. 15425102
Brand: Motic™ 12180142
Only null left
Add to basket
Add to basket
Widen your field of view for more complete specimen viewing. Motic™ Widefield Eyepiece WF10X/18mm provides an improved field of view and greater eye relief.
  • Widefield style provides improved field of view and greater eye relief.
  • Magnification: 10X.
  • Field of view: 18mm.
Specifications
Type Widefield
Description FOV: 18mm
For Use With (Equipment) RED100-Series, Series 18, Series 28, PM Series, DM1802-A Microscopes
Magnification Power 10X