Learn More
Motic™ Widefield Eyepiece N-WF10X/F.N.23 with Diopter Adjustment (ESD)
Widen your field of view for more complete specimen viewing. Motic™ Widefield Eyepiece N-WF10X/23mm with Diopter Adjustment (ESD) provides an improved field of view and greater eye relief.
Brand: Motic™ 1101001402711
Additional Details : Weight : 0.05000kg
Description
- Widefield style and diopter adjustment provides improved field of view and greater eye relief.
- Magnification: 10X.
- Field number: 23.
Specifications
Widefield | |
SM-171, GM Series Microscopes | |
10X |
F.N.: 23 | |
Diopter adjustment, ESD electrostatic shock protection |