Learn More
Wheaton™ White Urea Screw Caps
Shop All Wheaton Science Products Products
Click to view available options
Closure Size:
15.5 mm
22 mm
Unit Size:
Pack of 1000
Specifications
Specifications
| Closure Size | 22 mm |
| Liner Material | EPE |
| Description | Cap |
| Material | Urea |
| Color | White |
| Quantity | 1000 Pack |
| For Use With (Equipment) | Screw Thread Diagnostic Vials |