Learn More
Kautex™ White/Red Polypropylene Screw Closures
Product Code. 15231589
Click to view available options
Thread Size (Metric):
00 mm
Unit Size:
Pack of 100
Product Code. 15231589
Brand: Kautex™ 2000084201
Only null left
Specifications
Color White, Red
Material Polypropylene
Thread Size (Metric) 00 mm
Neck Finish 00 mm
Product Type Screw Cap
For Use With (Equipment) 10 to 1000 mL Bottle
Diameter (Metric) 32 mm