Learn More
Honeywell™ White Range Super Mont Blanc S2 Shoes
Click to view available options
Size:
38
39
40
42
44
47
Unit Size:
Each
Description
- Offers optimal protection
- White treated leather upper with limited stitching
- Resistant to liquid projections and easy to clean
Specifications
Specifications
| Certifications/Compliance | European Directive: 89/686/EEC - EC PPE Category II - Standard: EN 345-1 - Marking : S2 Quality Assurance: ISO 9001:2008 |
| Color | White |
| Material | Leather |
| Size | 38 |
| Sole Features | White, double density, injected, perforated |
| Sole Material | Polyurethane |
| Toe Style | Metallic 200J Toe Cap |
| Product Type | Safety Footwear |