Learn More
ABEBA™ White Microfiber ESD-Occupationa Loafer Shoes
Click to view available options
Size:
37
40
Unit Size:
1 pair
Specifications
Specifications
| Certifications/Compliance | CE, EN ISO 20347:2012, O1, SRA, DIN EN 61340 Standard |
| Color | White |
| For Use With (Application) | For healthcare and kitchen use |
| Liner Material | Silver Fibre |
| Material | Microfiber |
| Size | 37 |
| Sole Features | Shock-absorbent midsole |
| Sole Material | Polyurethane |
| Style | Occupationa Loafer |
| Toe Material | Steel |
| Show More |