Description
- Made from borosilicate glass
- Feature extra-heavy walls
Order Info
Stoppers and caps sold separately
Specifications
Specifications
| Color | Clear |
| Diameter (Metric) Outer | 17 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 4 mL |
| For Use With (Application) | For lyophilization and freeze drying applications |
| Volume (English) | 1.08 dr. |
| Height (Metric) | 43 mm |