Learn More
DWK Life Sciences Wheaton™ Unlined Aluminum Center Disc Tear-Out Seals
GREENER_CHOICE
Product Code. 11718779 Shop All Wheaton Science Products Products
Click to view available options
Color:
Blue
Red
Closure Size:
13 mm
20 mm
Unit Size:
Pack of 1000
Product Code. 11718779
Brand: DWK Life Sciences 22418205
Only null left
Add to basket
Add to basket
Exposes the top of the rubber stopper to the syringe when the center disc is opened or removed
  • Available in 13 or 20mm O.D.
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material Aluminum
Quantity 1000/Cs.
Type Center Disc Tear-out Cap
Closure Size 13 mm