Learn More
DWK Life Sciences Wheaton™ Snap-on Style Stoppers for Vials and Serum Bottles
Product Code. p-4607749 Shop All Wheaton Science Products Products
Click to view available options
Diameter (Metric) Inner:
3cm
Material:
Bromobutyl Rubber
Hardness:
50 Durometer
Unit Size:
Pack of 1000
Product Code. 13108720
Brand: DWK Life Sciences W224100342
Only null left
Add to basket
Add to basket
For maximum sealing of vials with blow-back glass finish
  • Made of gray bromobutyl
Not recommended for vials with a straight-walled finish.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 3cm
For Use With (Equipment) Fits 1.5cm I.D., 3cm O.D. mouth of vials and serum bottles
Hardness 50 Durometer
Autoclavable Autoclavable
Color Gray
Material Bromobutyl Rubber
Product Type Plug Stopper