Learn More
DWK Life Sciences Wheaton™ Safety-Coated Narrow Mouth Amber Bottles with Aluminum-Foil-Lined Black Phenolic Caps
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Closure Size:
28-400 mm
33-400 mm
Unit Size:
Pack of 12
Pack of 24
Description
- Plastisol-coated soda-lime glass
- Coating is inert and highly resistant to strong acids, alkalis, and most solvents
- Coating reduces risk of personal injury or contamination
- Black, foil-lined caps included
- Not autoclavable
Compliance
Helps meet OSHA requirements for safe handling and disposal of toxic or noxious substances.
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Capacity (English) | 32 oz. |
| Coating | Plastisol |
| Color | Amber |
| Diameter (English) | 3.7 in. |
| Material | Plastisol-coated Soda Lime Glass |
| Closure Color | Black |
| Certifications/Compliance | USP Type III |
| Closure Included | Yes |
| Height (Metric) | 210 mm |
| Show More |