Learn More
DWK Life Sciences Wheaton™ M-T Vial File™ and Accessories
Product Code. p-3207814 Shop All Wheaton Science Products Products
Click to view available options
Product Type:
Index Card
M-T Vial File
Partition
Storage Box
Quantity:
35 Case
6 Case
For Use With (Equipment):
228778 Hinged Case
228780 Hinged Case
24 x 20mL vials
40 x 4mL vials
54 x 8mL vials
60 x 2mL vials
W228790 Hinged Case
Unit Size:
Pack of 35
Pack of 6
Product Code. 11718719
Brand: DWK Life Sciences 228778
For compact and organized sample storage
  • Stores vials in clear polystyrene case
  • Foam inserts and partitions provide protection
  • With alphanumerical indexing card for identification
  • Index cards and partitions also available separately
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polystyrene
For Use With (Application) Vial storage and transport
For Use With (Equipment) 60 x 2mL vials
Includes Clear polystyrene hinged case, partitions, foam inserts and alphanumerical indexing card
No. of Places 60
Product Type M-T Vial File
Quantity 6 Case