Learn More
DWK Life Sciences Wheaton™ Jars with Polypropylene Screw Cap
Product Code. 12427391 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Autoclavable:
Autoclavable (Bottle Only)
Not Autoclavable
Color:
Clear
Natural
White
Unit Size:
Pack of 24
Pack of 36
Pack of 48
Pack of 72
Product Code. 12427391
Brand: DWK Life Sciences W209911
Only null left
Add to basket
Add to basket
  • Durable container with good chemical resistance
  • Pre-attached white polypropylene screw cap unlined or with foamed polyethylene liner
  • Polypropylene container is autoclavable; cap is not autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Natural
Diameter (English) 4.56 in.
Includes Screw Cap
Material Polypropylene
Closure Color White
Clarity Natural
Mouth Wide
Shape Round
Height (Metric) 97 mm
Product Type Jar
Height (English) 3.8 in.
Closure Liner Polyethylene
Closure Material Polypropylene
Closure Size 120-400 mm
Diameter (Metric) 116 mm
Closure Type Screw Cap