Learn More
DWK Life Sciences Wheaton™ E-Z Crimpers
Product Code. p-3205974 Shop All Wheaton Science Products Products
Click to view available options
For Use With (Equipment):
13 mm Flip-cap Seals
20 mm Flip-cap Seals
30 mm Aluminum Seals
Unit Size:
Each
Product Code. 10712573
Brand: DWK Life Sciences 224307
Only null left
Add to basket
Add to basket
Attach seals easily with these high-quality crimpers
  • Labeled for quick identification
  • Cushioned, short ergonomic handle
  • Polished crimping jaws
  • Spring assembly
  • Adjustable stop
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 30 mm Aluminum Seals