Learn More
DWK Life Sciences Wheaton™ E-Z Crimpers
Attach seals easily with these high-quality crimpers
Brand:  DWK Life Sciences 224323
631.75 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 0.57150kg
 View more versions of this product
Product Code. 10276284
  • 665.00 EUR / Each
Estimated Shipment: 30-05-2024
Description
  • Labeled for quick identification
  • Cushioned, short ergonomic handle
  • Polished crimping jaws
  • Spring assembly
  • Adjustable stop
Specifications
Specifications
Not Autoclavable
20 mm Flip-cap Seals