Learn More
DWK Life Sciences Wheaton™ Clear Boston Rounds with PE Cone-Lined Caps
Product Code. 12319739 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Closure Size:
20-400 mm
22-400 mm
24-400 mm
28-400 mm
33-400 mm
Unit Size:
Pack of 12
Pack of 24
Pack of 48
Product Code. 12319739
Brand: DWK Life Sciences W216816
Only null left
Add to basket
Add to basket
Ideal for solvent, chemical and sample storage
  • Well suited for storage and sampling of liquids; clear Type III soda-lime glass provides visibility of contents
  • Narrow mouth enables liquids to be carefully poured to prevent spillage
Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage.
Order Info
Caps can be ordered separately.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 16 oz., 500 mL
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Clear
Diameter (English) 2.95 in.
Material Soda Lime Glass
Closure Color Black
Certifications/Compliance EPA, FDA
Height (Metric) 168 mm
Height (English) 6.6 in.
Closure Liner Polyethylene
Closure Material Phenolic
Closure Size 28-400 mm
Diameter (Metric) 75 mm
Closure Type Screw Cap