Learn More
DWK Life Sciences Wheaton™ Amber Boston Rounds with Rubber-lined Caps
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
Closure Size:
20-400 mm
22-400 mm
24-400 mm
28-400 mm
33-400 mm
Unit Size:
Pack of 12
Pack of 24
Description
- Type III soda-lime glass
Specifications
Specifications
| Capacity | 2 oz., 60 mL |
| Capacity (Metric) | 60 mL |
| Capacity (English) | 2 oz. |
| Color | Amber |
| Diameter (English) | 1.22 in. |
| Material | Glass |
| Closure Color | Black |
| Height (Metric) | 79 mm |
| Height (English) | 3.1 in. |
| Closure Liner | Rubber |
| Show More |