Learn More
DWK Life Sciences Wheaton™ Amber Boston Rounds with Rubber-lined Caps
Product Code. 12399769 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
Closure Size:
20-400 mm
22-400 mm
24-400 mm
28-400 mm
33-400 mm
Unit Size:
Pack of 12
Pack of 24
Product Code. 12399769
Brand: DWK Life Sciences W216856
Only null left
Add to basket
Add to basket
Amber glass protects light-sensitive solutions
  • Type III soda-lime glass
TRUSTED_SUSTAINABILITY
Specifications
Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Amber
Diameter (English) 2.36 in.
Material Glass
Closure Color Black
Height (Metric) 138 mm
Height (English) 5.43 in.
Closure Liner Rubber
Closure Material Phenolic
Closure Size 24-400 mm
Diameter (Metric) 60 mm
Closure Type Screw Cap