Learn More
DWK Life Sciences Wheaton™ 12 x 32 mm ABC Clear Vial Pack (9 mm Screw Thread Finish and Marking Patch)
Product Code. 12444841 Shop All Wheaton Science Products Products
Click to view available options
Closure Color:
Black
Green
Natural
Septum:
PTFE
PTFE, Silicone
PTFE, Silicone, PTFE
Pre-slit:
No
Yes
Unit Size:
Pack of 100
Product Code. 12444841
Brand: DWK Life Sciences W2251510301
Only null left
Add to basket
Add to basket
ABC Vials, made from Type I borosilicate glass, and 9 mm ABC Screw Caps are available together in convenience packs of 100. The 1.8 mL vials are packaged in a recyclable PETE clamshell case to help prevent particulate contamination. Caps are available in six colors.
  • Manufactured from clear Type I borosilicate glass with writing patch
  • Packaged with 9 mm ABC Screw Caps
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Black
Color Clear
Diameter (Metric) Outer 12 mm
Material Borosilicate Glass
Pre-slit No
Volume (Metric) 1.8 mL
Septum PTFE, Silicone, PTFE
Height (English) 1.25 in.
Closure Material Polypropylene
Closure Type Screw Cap
Height (Metric) 32 mm
Quantity 100 Pk.
Closure Size 9 mm
Marking Spot Yes