Learn More
Cytiva Whatman™ Uniflo Syringe Filters, 0.45μm
Uniflo Syringe Filters
805.00 EUR - 855.00 EUR
Specifications
Pore Size | 0.45 μm |
---|---|
Sterility | Non-sterile |
Filtration Area | 0.65 cm2 |
Housing Material | Polypropylene |
Inlet Style | Femal Luer lock |
Product Code | Brand | Diameter (Metric) | Dimensions (L x W x H) | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Dimensions (L x W x H) | Material | Price | Quantity & Availability | |||
15843587
|
Cytiva
9910-1304 |
13 mm | 19.6 × 16.9 mm | Nylon |
855.00 EUR
Pack of 500 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||
15863587
|
Cytiva
9910-2504 |
25 mm | 24.5 × 29.2 mm | Nylon |
805.00 EUR
Pack of 500 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||
15803597
|
Cytiva
9911-2504 |
25 mm | 24.5 × 29.2 mm | PTFE |
805.00 EUR
Pack of 500 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||
15883587
|
Cytiva
9911-1304 |
13 mm | 19.6 × 16.9 mm | PTFE |
855.00 EUR
Pack of 500 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||
Description
PTFE membrane filters come in 0.22 μm and 0.45 μm pore sizes. Nylon is resistant to a range of organic solvents and suitable for use with high pH samples. Nylon may bind proteins, which makes it unsuitable for protein recovery applications.
Whatman Uniflo syringe filters from Cytiva’s Life Sciences business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of small volume samples up to 100 ml in laboratory sample prep. They are available in a variety of membrane choices and pore sizes with a polypropylene overmold housing. Uniflo filters are individually inkjet printed for easy filter identification and packaged in clean resealable, recyclable plastic boxs to optimize laboratory storage space.Specifications
0.45 μm | |
0.65 cm2 | |
Femal Luer lock | |
67.5 psi |
Non-sterile | |
Polypropylene | |
Male Slip Luer | |
500 Pieces |