Learn More
Cytiva Whatman™ Nuclepore™ Polycarbonate Hydrophilic Membrane Sheets
Product Code. p-112308
Change view
Click to view available options
Pore Size:
0.03 μm
0.05 μm
Dimensions (L x W):
42 x 19mm
254 x 204mm
Unit Size:
Pack of 100
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Dimensions (L x W) unitSize
11329004 0.03 μm 254 x 204mm Pack of 25
11362915 0.05 μm 42 x 19mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11329004 Supplier Cytiva Supplier No. 113502
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from high-quality polycarbonate film and track-etched for high pore size precision. Whatman™ Nuclepore™ Polycarbonate Hydrophilic Membrane Sheets combine high flow rates and chemical and thermal resistance with sharply defined pores, making them suitable for microscopy, cytology, parasitology, and certain air and oceanography analyses.
Effective chemotaxis requires a delicate balance between cell adhesion and migration. Nucleopore Membranes combine PVP-free polypropylene with sharp, uniform cylindrical pores to achieve both.
  • Offers a highly defined pore size cutoff in the sub-micron range, enabling particle deformity measurement and capture of extremely small particles
  • Track-etched for highly defined pore size and high pore density
  • Treated with PVP to render it hydrophilic
  • Low extractables, minimizing sample contamination risk
  • High chemical resistance and good thermal stability for a wide range of sample types
  • Low, consistent ash and low tare weights
  • Smooth flat surface for high particle visibility
TRUSTED_SUSTAINABILITY
Specifications
Material Nucleopore Polycarbonate
Pore Size 0.03 μm
Flow Rate High
Hydrophobicity Hydrophilic
Type Membrane Filter
Dimensions (L x W) 254 x 204mm