Learn More
QIAGEN QIAcard FTA Wash Buffer
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
For contamination-free collection, storage and transport of DNA samples.
QIAcard FTA Wash Buffer is used for purifying nucleic acids stored on FTA cards.
Specifications
Specifications
| For Use With (Equipment) | QIAcard FTA cards, indicating FTA cards and EasiCollect device |
| For Use With (Application) | Rapid purification of DNA from blood, cells, tissue, yeast, bacteria |
| Quantity | 500 mL |