Learn More
QIAGEN QIAcard FTA CloneSaver
Click to view available options
Quantity:
1 Pk.
Unit Size:
Pack of 5
Description
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. 96 sample areas per card.
CloneSaver Card is designed for the collection, long-term storage and purification of plasmid and BAC DNA from bacterial clones in a 96-well format.
Specifications
Specifications
| Description | Designed for the collection, long-term storage, and purification of plasmid and BAC DNA from bacterial clones in a 96-well format. |
| Format | 96-well Plate |
| For Use With (Application) | Designed for the collection, long-term storage, and purification of plasmid and BAC DNA from bacterial clones in a 96 well format. |
| Product Type | CloneSaver Card |
| Quantity | 1 Pk. |
| Sample Volume | 5 mL |
Safety and Handling
Signal Word
- Warning