Learn More
Cytiva Whatman™ Drying Rack for 903 Protein Saver Cards
Description
Air-dries blood specimens in a suspended horizontal position. Whatman™ Drying Rack for 903 Protein Saver Card is easily assembled and can be affixed to a wall or counter top with optional hook-and-loop stickers.
- For use with neonatal human diagnostic testing cards
- CE Marked
- Accommodate multiple collection forms at one time
Specifications
Specifications
| For Use With (Equipment) | 903 Protein Saver Card |
| Quantity | 10 Pack |
| Includes | Mounting Sticker not included |
| Product Type | Drying Rack |