Learn More
Cytiva Whatman Cyclopore Polycarbonate Membrane
Description
- Low affinity for stains improves visibility under microscope
- Low protein binding minimizes sample loss of proteinaceous samples
- Narrow pore size distribution and high pore density support rapid surface capture of analyte
- Negligible absorption and adsorption of filtrate; nonhygroscopic
- No particle shedding enables clean filtrate
- Biologically inert
Specifications
Specifications
| Material | Polycarbonate |
| Pore Size | 5 μm |
| For Use With (Application) | Epifluorescence Microscopy, Environmental Analysis, Cell Biology, EPA Testing, Fuel Testing, Bioassays, Parasitology, Air Analysis, Water Microbiology |
| Quantity | 100 Pk. |
| Type | Membrane Circle |
| Dimensions (L x W) | 25 x 80 mm |