Learn More
Honeywell Safety Products™ Welder Gloves
Product Code. 12641003
Change view
Click to view available options
:
Size: 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12641003 Size: 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12641003 Supplier Honeywell Safety Products™ Supplier No. 201280110
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Especially suitable for medium handling and light welding, these gloves are constructed of heat-resistant grain leather. Honeywell Safety Products™ Welder Gloves are designed with a split leather cuff to protect a large part of the forearm.
  • Suitable for light welding.
  • Heat-resistant cowhide grain leather construction.
  • Gunn cut, sewn-on thumb, Crispin type.
  • Split leather cuff to protect the forearm.
  • U.S. assembly.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 407-2004 Heat hazards, EC Category PPE 2, EC Attestation Number 0075/797/162/01/07/0005
Classification Dexterity: 5
Cuff Material Split Leather
Cut Style Gunn
Cut Resistance Rating 1
Material Grain Leather
Product Type Welding Gloves
Size 10
Style Gauntlet
Thickness (Metric) 0.9 to 1 mm
For Use With (Application) Welding
Puncture Rating 1
Quantity 10Pair/PK
Resistant To Heat
Tear Rating 2
Length (Metric) Cuff 15 cm