Learn More
Trajan™ Waters WISP Syringe
HPLC Autosampler syringe with 1/4-28 UNF screw thread
185.00 EUR
Specifications
Product Type | Replacement Plunger |
---|---|
For Use With (Equipment) | Waters Intelligent Sample Processor Syringe |
Product Code | Brand | Volume (Metric) | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Model | Price | Quantity & Availability | ||||
11907234
|
Trajan™
003990 |
25 μL | 25D-WISP |
185.00 EUR
Each |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Waters instruments
Specifications
Replacement Plunger | |
Waters Intelligent Sample Processor Syringe |