Learn More
Trajan™ Waters WISP Syringe
HPLC Autosampler syringe with 1/4-28 UNF screw thread
Brand:  Trajan™ 003990
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11907234
  • 185.00 EUR / Each
Estimated Shipment: 28-06-2024
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Waters instruments
Specifications
Specifications
Replacement Plunger
25D-WISP
25 μL
Waters Intelligent Sample Processor Syringe