Learn More
Palintest™ Water Testing Tablet
Description
- Ideal for quick and reliable testing in the field
- They are crushed in the water sample until a color change occurs and the number required allows results to be calculated in ppm or mg/L
- Magnesium hardness is the difference between total and calcium hardness
- Permanent hardness is the difference between total hardness and total alkalinity
- In bottles of 250 tablets with instructions
Specifications
Specifications
| For Use With (Application) | Total Alkalinity |
| Quantity | 250 Pack |
| Type | Water Testing Tablet |