Learn More
Thermo Scientific™ Water Purification Systems Storage Reservoirs
Product Code. 11397091 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Feed Water Tank
Sterile Overflow
Storage Reservoir
Capacity (Metric):
100 L
30 L
30 to 100 L
60 L
Includes:
Level Display
Level Display, Recirculation Pump
N/A
Reservoir
Tank
For Use With (Equipment):
GenPure, GenPure Pro, GenPure xCAD
Pacific RO, Pacific TII
Pacific TII
Smart2Pure
Smart2Pure 12
Smart2Pure, Pacific and LabTower Water Purification Systems
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Capacity (Metric) Includes For Use With (Equipment) unitSize
11397091 Storage Reservoir 100 L Level Display Pacific RO, Pacific TII Each
11377081 Storage Reservoir 30 L Level Display Pacific RO, Pacific TII Each
11367061 Sterile Overflow 30 to 100 L N/A Smart2Pure, Pacific and LabTower Water Purification Systems Each
13498709 Sterile Overflow 30 to 100 L N/A Smart2Pure Each
11327091 Storage Reservoir 60 L Level Display, Recirculation Pump Pacific TII Each
11367081 Storage Reservoir 30 L Level Display, Recirculation Pump Pacific TII Each
11317091 Storage Reservoir 30 L Reservoir Smart2Pure 12 Each
11307091 Feed Water Tank 30 L Tank GenPure, GenPure Pro, GenPure xCAD Each
11337091 Storage Reservoir 60 L Level Display Pacific RO, Pacific TII Each
11387091 Storage Reservoir 100 L Level Display, Recirculation Pump Pacific TII Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11397091 Supplier Thermo Scientific™ Supplier No. 50155503
Only null left
Add to basket
Add to basket
Safely store purified water from your Thermo Scientific™ Barnstead™ GenPure™, Pacific™ or Smart2Pure™ system with one of these compatible storage reservoirs.
Safely store purified water from a compatible system with Thermo Scientific™ Water Purification Systems Storage Reservoirs. For use with Thermo Scientific™ Barnstead™ GenPure™, Pacific™ or Smart2Pure™ systems, these reservoirs are compatible with the following models: GenPure, GenPure Pro, GenPure xCAD, GenPure xCAD Plus, Pacific RO, Pacific TII and Smart2Pure.
Features:
  • Large opening offers easy and effective cleaning.
  • Optional safety filters include a CO2 absorber prevents conductivity increases from drawn-in CO2 and a sterile overflow to prevent microorganisms from entering and contaminating reservoir water.
  • Polyethylene reservoir is constructed with inert materials opaque to light.
  • Recirculation pump (optional) protects water from bacterial growth during standstill while maintaining a low conductivity value.
  • Wall mount is optional.
  • Conical bottom outlet allows for complete draining.
For use with GenPure, GenPure Pro, and GenPure xCAD purification systems only:
  • 06.5038
  • 06.5068
For use with Pacific RO water purification systems only:
  • 06.5034
  • 06.5064
  • 06.5084
For use with Pacific TII water purification systems only:
  • 06.5032
  • 06.5062
  • 06.5082
For use with Pacific RO & Pacific TII water purification systems only:
  • 06.5033
  • 06.5063
  • 06.5083
For use with Smart2Pure 12 water purification systems only:
  • 06.5040
  • 06.5070
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 L
For Use With (Equipment) Pacific RO, Pacific TII
Type Storage Reservoir
Includes Level Display