missing translation for 'onlineSavingsMsg'
Learn More

Water with 0.1% Formic Acid, LC-MS Grade, Scharlab

Product Code. 11322625
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11322625 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11322625 Supplier Scharlab Supplier No. AG00081000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for use as an eluent in LC-MS. Water with 0.1% Formic Acid, LC-MS Grade, Scharlab is free from alkaline impurities that can lead to the rise of ions that increase background noise and cause interference in the spectrum.

LC-MS is a powerful technique combining separation by HPLC with the structural information obtained by MS. Its application is widespread in the scientific and pharmaceutical fields as well as in biotechnology and pesticide residue analysis.

  • Designed with particularly low content (<100 ppb) of alkaline metals, especially K and Na
  • Microfiltered through a 0.22μm filter to eliminate any solid particles
  • Quality of the eluents is ensured by means of suitability tests for LC-MS
  • To improve ion generation, the addition of a volatile organic acid to the mobile phase is recommended

Specifications

Absorbance 1.301 at 210nm, 0.347 at 230nm, 0.004 at 254nm
pH 2.6 - 2.8
CAS 7732-18-5, 64-18-6
Molecular Formula H2O
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Miscible in water
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Grade Microfiltered through membranes of pore diameter 0.22μm
Concentration or Composition (by Analyte or Components) 0.1% Formic acid
Density 1.00g/cm3
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.