Learn More
Ohaus™ Wall Mount Clamp
Shop All Ohaus Products
Click to view available options
Length (Metric):
80.00 mm
Unit Size:
Each
Description
- Designed to securely grip and position laboratory apparatus
- Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
- Fixed position
- Dual adjust
Specifications
Specifications
| Type | Wall Mount Clamp |
| Material | Nickel-plated |
| Description | Clamp, Specialty, Wall, CLS-WALLCZ |
| Length (English) | 3.15 in. |
| Length (Metric) | 80.00 mm |
| Size (Metric) Grip | 5 to 10 mm |
| Jaw Type | 2-Prong |