Learn More
SI Analytics Wetting cap for 12mm electrodes
Brand:  SI Analytics 285123185
Wetting cap for 12mm electrodes
Product Code. 10009890
  • ONLINE EXCLUSIVE PRICE
    7.45 EUR
    6.50 EUR / Each
    Save 0.95 EUR
    12% Off
Only null left
Estimated Shipment: 12-02-2026