missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VU0357017 hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
VU0357017 hydrochloride (CID-25010775) is a potent, selective and brain-penetrant allosteric agonist of M1 muscarinic acetylcholine receptor, with an EC50 of 477 nM. VU0357017 hydrochloride is highly selective for M1 and has no activity at M2-M5 up to the highest concentrations tested (30 μM). VU0357017 hydrochloride can be used for the research of Alzheimer’s disease and schizophrenia.
Specifications
Specifications
| Chemical Name or Material | VU0357017 hydrochloride |
| CAS | 1135242-13-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H28ClN3O3 |
| Quantity | 100 mg |
| Synonym | CID-25010775 |
| Solubility Information | H2O : 33.33 mg/mL (90.11 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (67.59 mM; Need ultrasonic) |
| SMILES | O=C(N1CCC(NCCNC(C2=CC=CC=C2C)=O)CC1)OCC.[H]Cl |
| Molecular Weight (g/mol) | 369.89 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction