missing translation for 'onlineSavingsMsg'
Learn More

VU 0240551, Tocris Bioscience™

Product Code. p-4817240
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18095112

Brand: Tocris Bioscience 3888/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

KCC2 inhibitor

Inhibitor of the neuronal K-Cl cotransporter, KCC2 (IC50 = 560 nM for K+ uptake assay in KCC2-overexpressing cells). Exhibits selectivity over the Na-K-2Cl cotransporter, NKCCl. Also inhibits hERG and L-type Ca2+ channels.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 893990-34-6
Molecular Formula C16H14N4OS2
Molecular Weight (g/mol) 342.435
InChI Key WJRWSLORVIHRNX-UHFFFAOYSA-N
Synonym n-4-methyl-2-thiazolyl-2-6-phenyl-3-pyridazinyl thio-acetamide, n-4-methyl-2-thiazolyl-2-6-phenyl-3-pyridazinyl thio acetamide, n-4-methyl-1,3-thiazol-2-yl-2-6-phenylpyridazin-3-yl thio acetamide, n-4-methylthiazol-2-yl-2-6-phenylpyridazin-3-ylthio acetamide, n-4-methyl-1,3-thiazol-2-yl-2-6-phenylpyridazin-3-yl sulfanylacetamide, n-4-methyl-1,3-thiazol-2-yl-2-6-phenylpyridazin-3-yl sulfanyl acetamide, d0m2pp, cid, vu hplc, n-4-methylthiazol-2-yl-2-6-phenylpyridazin-3-yl thio acetamide
PubChem CID 7211972
IUPAC Name N-(4-methyl-1,3-thiazol-2-yl)-2-(6-phenylpyridazin-3-yl)sulfanylacetamide
SMILES CC1=CSC(=N1)NC(=O)CSC2=NN=C(C=C2)C3=CC=CC=C3

Specifications

Quantity 10 mg
Formula Weight 342.43
Percent Purity >99%
Chemical Name or Material VU 0240551
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.