Learn More
VTP-27999 TFA, MedChemExpress
Product Code. 16407876
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16407876
Brand: Medchem Express HY50769
MedChemExpress VTP-27999 TFA is an alkyl amine Renin inhibitor; VTP-27999 TFA is useful for Hypertension and End-Organ Diseases.
VTP-27999 TFA is an alkyl amine Renin inhibitor
Specifications
Chemical Name or Material VTP-27999 TFA
Physical Form Oil
CAS 1013937-63-7
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C28H42ClF3N4O7
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 10 mg/mL (15.65 mM; Need ultrasonic) ∣Ethanol : 2.4 mg/mL (3.76 mM; Need ultrasonic)
SMILES O=C(O)C(F)(F)F.COC(NCCO[C@@H](C1=CC(Cl)=CC=C1)[C@@]2([H])CN(CCC2)C(NC[C@@H](NC)C[C@H]3CCCOC3)=O)=O
Molecular Weight (g/mol) 639.1
Formula Weight 639.1
Percent Purity 97.2%
Grade Research