Learn More
IKA™ VSE 1 Vacuum Safety Emmission
Product Code. 16218711 Shop All IKA Products
Change view
Click to view available options
Weight (Metric):
1.145 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) unitSize
16218711 1.145 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16218711 Supplier IKA™ Supplier No. 0035001873
Only null left
Add to basket
Add to basket
Designed for use with a range of rotary evaporators.
  • For use with the following IKA™ models:
    • RV 3 V, RV 3 V-C, Vacstar Digital, RV 3 FLEX, RV 3 with dry ice condenser (coated), RV 8 V, RV 8 V-C, RV 8 FLEX, RV 10 Auto V, RV 10 Auto V-C, RV 10 Auto FLEX
TRUSTED_SUSTAINABILITY
Specifications
Weight (Metric) 1.145 kg