missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS13B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24505-1-AP-20UL
This item is not returnable.
View return policy
Description
COH1 (Cohen syndrome protein 1), also known as VPS13B (vacuolar protein sorting-associated protein 13B) or CHS1, belongs to the VPS13 family and may function in vesicle-mediated transport and sorting of proteins within the cell. COH1 is widely expressed and multiple alternatively spliced transcript variants have been observed. Mutations in this gene have been associated with Cohen syndrome. COH1 is a Golgi-localized peripheral membrane protein and plays a critical role in Golgi (re)assembly.Specifications
| VPS13B | |
| Polyclonal | |
| Unconjugated | |
| VPS13B | |
| CHS1, COH1, Cohen syndrome protein 1, KIAA0532, VPS13B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 157680, 666173 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7Z7G8, Q80TY5 | |
| VPS13B | |
| VPS13B Fusion Protein Ag21368 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction