Learn More
VPS13B Monoclonal Antibody (OTI6F6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807801
This item is not returnable.
View return policy
Description
COH1 (Cohen syndrome protein 1), also known as VPS13B (vacuolar protein sorting-associated protein 13B) or CHS1, belongs to the VPS13 family and may function in vesicle-mediated transport and sorting of proteins within the cell. COH1 is widely expressed and multiple alternatively spliced transcript variants have been observed. Mutations in this gene have been associated with Cohen syndrome. COH1 is a Golgi-localized peripheral membrane protein and plays a critical role in Golgi (re)assembly.Specifications
| VPS13B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7Z7G8 | |
| VPS13B | |
| Human recombit protein fragment corresponding to amino acids 91-346 of human VPS13B produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI6F6 | |
| Unconjugated | |
| VPS13B | |
| CHS1, COH1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 157680 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |