Learn More
VPS13B Monoclonal Antibody (OTI6F6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA807801
Encompass_Preferred
 View more versions of this product
Product Code. 17558603
  • 540.87 EUR / 100µL
Description
COH1 (Cohen syndrome protein 1), also known as VPS13B (vacuolar protein sorting-associated protein 13B) or CHS1, belongs to the VPS13 family and may function in vesicle-mediated transport and sorting of proteins within the cell. COH1 is widely expressed and multiple alternatively spliced transcript variants have been observed. Mutations in this gene have been associated with Cohen syndrome. COH1 is a Golgi-localized peripheral membrane protein and plays a critical role in Golgi (re)assembly.
Specifications
Specifications
VPS13B
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q7Z7G8
VPS13B
Human recombit protein fragment corresponding to amino acids 91-346 of human VPS13B produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI6F6
Unconjugated
VPS13B
CHS1, COH1
Mouse
Affinity Chromatography
RUO
157680
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions