Learn More
Kartell SpA Volumetric Flask with Screw Cap
Product Code. 10308661
Change view
Click to view available options
Capacity (Metric):
100 mL
500 mL
1000 mL
Unit Size:
Pack of 10
Pack of 20
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10308661 100 mL Pack of 20
10114671 500 mL Pack of 10
10073021 1000 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10308661 Supplier Kartell SpA Supplier No. 0249100
Only null left
Add to basket
Add to basket
Volumetric flask with cap, PMP
Crystal clear volumetric flasks create no meniscus so final readings are easy. Calibrated gravimetrically at 20°C. Thick walls eliminate distortion. Thin necks make final measurement precise. Autoclavable to 170°C. Chemically nonabsorbant. No wetting. Screw caps included.
Specifications
Capacity (Metric) 100 mL
Material PMP
Product Type Volumetric Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Threaded
Sterility Non-sterile
Bottom Shape Round
Surface Treatment Untreated