Learn More
Kartell SpA Volumetric Flask with Screw Cap
Description
Crystal clear volumetric flasks create no meniscus so final readings are easy. Calibrated gravimetrically at 20°C. Thick walls eliminate distortion. Thin necks make final measurement precise. Autoclavable to 170°C. Chemically nonabsorbant. No wetting. Screw caps included.
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Material | PMP |
| Product Type | Volumetric Flask |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | No |
| Mouth Style | Threaded |
| Sterility | Non-sterile |
| Bottom Shape | Round |
| Surface Treatment | Untreated |