Learn More
Karl Hecht™ Volumetric Flask with PP Stopper, Conformity Certified
Product Code. 10490172
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
10 L
Closure Size:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 29/32
NS 34/35
NS 45/40
NS 7/16
Tolerance:
0.025 mL
0.04 mL
0.06 mL
0.10 mL
0.15 mL
0.25 mL
0.4 mL
0.6 mL
1.2 mL
2.0 mL
Neck Style:
Standard
Wide
Unit Size:
Each
Product Code. 10490172
Brand: Karl Hecht™ 42096028
Only null left
Add to basket
Add to basket
Simply and accurately measure required sample volumes. Karl Hecht™ Volumetric Flask with PP Stopper Conformity Certified is thick-walled, clearly marked and labeled, and has a solid finish
  • Duran™ borosilicate glass 3.3.
  • Standard type.
  • Standard ground and interchangeable NS stopper of polyethylene.
  • Fused blue graduation.
  • Thick-walled, solid finish.
  • Clearly legible figures and marks.
  • Conformity certified.
    • Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications
Certifications/Compliance DIN EN ISO 1042
Closure Material Polyethylene
Capacity (Metric) 25 mL
Material Borosilicate Glass (3.3)
Neck Style Standard
Product Type Volumetric Flask
Tolerance 0.04 mL
Graduated Yes
Closure Type NS Stopper
Closure Size NS 10/19