Learn More
Karl Hecht™ Volumetric Flask with PP Stopper, Conformity Certified
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
10 L
Closure Size:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 29/32
NS 34/35
NS 45/40
NS 7/16
Tolerance:
0.025 mL
0.04 mL
0.06 mL
0.10 mL
0.15 mL
0.25 mL
0.4 mL
0.6 mL
1.2 mL
2.0 mL
Unit Size:
Each
Neck Style:
Standard
Wide
Description
- Duran™ borosilicate glass 3.3.
- Standard type.
- Standard ground and interchangeable NS stopper of polyethylene.
- Fused blue graduation.
- Thick-walled, solid finish.
- Clearly legible figures and marks.
- Conformity certified.
- Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications
Specifications
| Closure Material | Polyethylene |
| Capacity (Metric) | 2 mL |
| Material | Borosilicate Glass |
| Shape | Conical |
| Tolerance | 0.025 mL |
| Bottom Shape | Conical |
| Closure Type | NS Stopper |
| Closure Size | NS 7/16 |