Learn More
Karl Hecht™ Volumetric Flask with PP Stopper, Conformity Certified
Product Code. 10083891
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
10 L
Closure Size:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 29/32
NS 34/35
NS 45/40
NS 7/16
Tolerance:
0.025 mL
0.04 mL
0.06 mL
0.10 mL
0.15 mL
0.25 mL
0.4 mL
0.6 mL
1.2 mL
2.0 mL
Neck Style:
Standard
Wide
Unit Size:
Each
Product Code. 10083891
Brand: Karl Hecht™ 42096096
Only null left
Add to basket
Add to basket
Simply and accurately measure required sample volumes. Karl Hecht™ Volumetric Flask with PP Stopper Conformity Certified is thick-walled, clearly marked and labeled, and has a solid finish
  • Duran™ borosilicate glass 3.3.
  • Standard type.
  • Standard ground and interchangeable NS stopper of polyethylene.
  • Fused blue graduation.
  • Thick-walled, solid finish.
  • Clearly legible figures and marks.
  • Conformity certified.
    • Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications
Certifications/Compliance DIN EN ISO 1042
Closure Material Polyethylene
Capacity (Metric) 5 L
Material Borosilicate Glass
Neck Style Standard
Product Type Volumetric Flask
Tolerance 1.2 mL
Graduated Yes
Closure Type NS Stopper
Closure Size NS 34/35