Learn More
Poulten Graf™ Volac Class A Borosilicate Glass Mixing Cylinder
Specifications
Specifications
| Base Type | Hexagonal |
| Scale Color | Blue |
| Shape | Round |
| Type | Graduated Mixing Cylinder |
| Material | Borosilicate Glass |
| Class | A |
| Capacity (Metric) | 100 mL |
| Includes | PP Stopper |
| Stopper Material | Polypropylene |
| Graduations | 1 mL |